Learn More
Charles River Laboratories ENDOSAFE PTS CARTIRIDGE 10/PK
Supplier: Charles River Laboratories PTS2005
Cartridge; Endosafe PTS; Charles River Endosafe; For endotoxin determination using the PTS platform; Sensitivity 0.05 EU/mL