Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Copan Diagnostics eNat™ 6U074S01
Minitip flocked swab and eNat™ medium
Supplier: Copan Diagnostics 6U074S01
Description
- 80 mm breakpoint
- Packaged with 2 mL eNat™ guanidine medium in skirted tube with plastic cap
- Individually packaged
- Sterile
Specifications
Minitip flocked swab and 2 mL eNat™ medium in skirted tube with plastic cap | |
Individually packaged | |
80 mm |
Sterile | |
Specimen collection and transport for molecular assays | |
eNat™ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction