Learn More
MilliporeSigma™ Supelco™ Empty Glass GC Column, for Varian™ 3000, 4000, 6000, and Vista (FID, Inj. A to Det. A, On-Column)
Empty column may be used for packing adsorbents for temperature-programmed desorption (TPD) instrumentation.
Supplier: MilliporeSigma™ Supelco™ 21207
Description
Silane treated empty glass column. Fittings are not included. This column fits Varian 3000, 4000, 6000, and Vista (FID, injector A to detector A, on-column injection)Specifications
Empty GC | |
Gas Chromatography (GC) | |
Glass Column |
2 mm | |
1 m | |
None |