Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDS Analytical™ Empore™ C18/SCX StageTips
C18 bonded silica and cation exchange (SCX) StageTips are SPE micropipette tips for the desalting of large peptides and proteins, as well as peptide fractionation.
$190.80 - $1422.00
Specifications
Particle Size | 12 μm |
---|---|
Product Line | Empore |
Sorbent | DVB (Polystyrene-divinylbenzene) |
Volume (Metric) | 200 μL |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
13-110-067
|
CDS Analytical™
70201910076EA |
96 Pk. |
Pack of 96 for $190.80
|
|
|||||
13-110-068
|
CDS Analytical™
70201910076 |
960 Cs. |
Case of 960 for $1,422.00
|
|
|||||
Description
CDS™ Empore™ StageTips C18 bonded silica and cation exchange (SCX) solid phase extraction micropipette tips combine good retention capacity with high recoveries.
- Recommended for desalting of large peptides and proteins and peptide fractionation at acidic, neutral, and basic pH conditions
Specifications
12 μm | |
Empore | |
DVB (Polystyrene-divinylbenzene) | |
200 μL |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title