Learn More
Cytiva Pallflex Emfab™ Air Monitoring Filters - TX40HI20WW, 47 mm (100/pkg)
Supplier: Cytiva 7221
Versatile Filters Uniquely Suited for a Broad Range of Air Monitoring ApplicationsUseful for high temperature and hot gas air monitoring applications.Ideal for stack sampling and diesel emissions testing. This product is not approved for use in medical, clinical, surgical or other patient applications.