Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ ZHE Hazardous Waste System
ZHE (Zero Head Space Extractor) system has been designated by the USEPA as a suitable apparatus for Toxicity Characteristic Leaching Procedure (TCLP).
Supplier: MilliporeSigma™ YT30090HW
Description
Cellulose support pads are used to reinforce filters in Millipore's monitors for contamination analysis. When saturated with growth medium, they can also be used for culturing microorganisms on membranes.To increase flow rate and throughput, our woven mesh spacers can be placed between filters in a serial filtration to prevent the downstream screen filter from in.blinding in. the upstream filter pores.
Features & Benefits
- Avoid membrane damage during high pressure or fast flow conditions
- Spacers enable you to combine multiple stages of filtration into a single process step, saving time
Applications: Air Monitoring, Environmental Monitoring, Aerosol Contamination Monitoring
Compliance
Designated by the U.S. EPA as a suitable apparatus for TCLP
Specifications
Not Autoclavable | |
316 Stainless Steel Body; Molded PP Handwheels; Fluoroelastomer-A O-Rings | |
3.5 bar | |
Environmental Test Kits or Supplies | |
15.9 cm |
Hazardous Waste System | |
Non-sterile | |
3.5 bar | |
27.9 cm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction