Learn More
MilliporeSigma™ Novagen™ Veggie NovaBlue Singles™ Competent Cells
Produces high-quality plasmid DNA
$302.00 - $537.00
Specifications
Efficiency Guaranteed efficiency: >1.5 x 108 cfu/μg
Content And Storage ≤-70°C
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
71-251-3
View Documents
MilliporeSigma™
712513
11 Reactions
Each for $302.00
Add to Cart
71-251-4
View Documents
MilliporeSigma™
712514
22 Reactions
Each for $537.00
Add to Cart
Description
Description
Veggie™ NovaBlue Singles™ Competent Cells are available for routine cloning applications. NovaBlue is a K-12 strain ideally suited as an initial cloning host due to its high transformation efficiency, blue/white screening capability (with appropriate plasmids), and mutations in recA and endA, which result in high yields of high-quality plasmid DNA.
Specifications
Specifications
Guaranteed efficiency: >1.5 x 108 cfu/μg
≤-70°C