Learn More
MilliporeSigma™ Novagen™ Rosetta™ Blue Competent Cells
RosettaBlue host strains are NovaBlue derivatives that combine high transformation efficiency and with enhanced expression of eukaryotic proteins that contain codons rarely used in E. coli.
$189.00 - $346.00
Specifications
Efficiency | Guaranteed efficiency: >1 x 106 cfu/μg |
---|---|
Content And Storage | ≤-70°C |
Description
RosettaBlue™ host strains are NovaBlue derivatives that combine high transformation efficiency and recA endA lacIq mutations with enhanced expression of eukaryotic proteins that contain codons rarely used in E. coli. These strains supply tRNAs for AGG, AGA, AUA, CUA, CCC, and GGA on a chloramphenicol-resistant plasmid. The tRNA genes are driven by their native promoters.Specifications
Guaranteed efficiency: >1 x 106 cfu/μg | |
≤-70°C |