Learn More
MilliporeSigma™ Novagen™ Origami™ B Competent Cells
Origami B host strains carry the same mutations as the original Origami strain, except that they are derived from a lacZY mutant of BL21 to enable precise control of expression levels using IPTG.
$202.00 - $330.00
Specifications
Efficiency | Guaranteed efficiency: >2 x 106 cfu/μg |
---|---|
Content And Storage | ≤-70°C |
Description
Origami™ B host strains carry the same trxB/gor mutations as the original Origami strains, except that they are derived from a lacZY mutant of BL21. Thus the Origami B strains combine the desirable characteristics of BL21, Tuner™ and Origami hosts in one strain background. The trxB and gor mutations are selectable on kanamycin and tetracycline, respectively; therefore, these strains are recommended for use only with pET plasmids carrying the ampicillin resistance marker bla.Specifications
Guaranteed efficiency: >2 x 106 cfu/μg | |
≤-70°C |