Learn More
MilliporeSigma™ Novagen™ His.Tag™ Antibody HRP Conjugate Kit
Horseradish peroxidase-conjugated His·Tag monoclonal antibody used as single detection reagent
Supplier:  MilliporeSigma™ 718403
Catalog No. 71-840-3MI
Add to Cart
Description
Description
  • Single detection reagent for His·Tag fusion proteins
  • Eliminates cross-reactivity associated with secondary antibody reagents
  • Produces strong signal at a 1:1000 to 1:2000 dilution on Western blots and dot blots
  • Antibody also detects marker bands in Novagen Perfect Protein™ Western Markers
  • Negligible cross-reactivity to bacterial, yeast, insect, or mammalian cell proteins
  • Contains sufficient reagents for 25 to 50 blots and includes 5% Alkali-soluble Casein for blocking reagent
Specifications
Specifications
125μL
+2° to +8°C
Product Suggestions
Product Suggestions