Learn More
MilliporeSigma™ MQuant™ Lead(II) Acetate Reagent Papers
Use for the determination of sulfide and hydrogen sulfide
Supplier:  MilliporeSigma™ 1.09511.0003
Catalog No. M1095110003
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Includes: Reagent
Description
Description
High-tech, fast and simple means for semiquantitative testing
  • Store at 15° to 25°C
  • Small and easy to handle for on-site use
  • Fast results
  • Ideal for screening
  • PET film and low reagent content for easier disposal
Specifications
Specifications
For the semiquantitative detection of ions and of inorganic and organic substances and can easily be used anywhere, lead (II) acetate paper is used for the determination of sulfide and hydrogen sulfide
3 Rolls
15°C to 25°C
Lead Acetate Reagent Paper
Product Suggestions
Product Suggestions