Learn More
MilliporeSigma™ Milli-Q™ Advantage A10 Water Purification Packs
For use with Advantage A10 system
$1100.00
Specifications
For Use With (Application) | Spectrophotometry, spectroscopy, chromatography, and biochemistry |
---|---|
For Use With (Equipment) | Milli-Q™ Advantage 10 water purification system |
Catalog Number | Mfr. No. | Type | Feedwater Source | Purification Method | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Feedwater Source | Purification Method | Price | Quantity & Availability | |||
QGARDT1X1
|
MilliporeSigma™
QGARDT1X1 |
Q-Gard T1 Purification Pack | Elix, Reverse Osmosis, Distilled | Ion Exchange, Activated Carbon |
Each for $1,100.00
|
|
|||
QGARDT2X1
|
MilliporeSigma™
QGARDT2X1 |
Q-Gard T2 Purification Pack | Deionized | Ion Exchange, Activated Carbon, Depth Filtration |
Each for $1,100.00
|
|
|||
Description
For use with Advantage A10 system
Spectrophotometry, spectroscopy, chromatography, and biochemistry
Certifications
ISO 9001 v.2000 and ISO 14001
Compliance
USP 645 suitability requirements
Specifications
Spectrophotometry, spectroscopy, chromatography, and biochemistry | |
Milli-Q™ Advantage 10 water purification system |