Learn More
MilliporeSigma™ MColortest™ Chlorine Test Kits
can be used for the determination of chlorine.
Manufacturer:  MilliporeSigma™ 1.14826.0001
 View more versions of this product
Catalog No. M1148260001
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
The liquid reagent can be used for the determination of free and total chlorine with the MColortestTM Chlorine test. The chlorine concentration can be measured semiquantitatively by visual comparison of the colour of the measurement solution with the colour fields of the colour disk.
Specifications
Specifications
0.25 to 5 mg/L Cl2
Chlorine Test Kit