Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ PANSORBIN™ Cells
For mitogenic stimulation of B lyphocytes and for immunoprecipitation
$130.00 - $2387.00
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-785-81GM
|
MilliporeSigma™
5078581GM |
1 g |
Each for $130.00
|
|
|||||
50-785-85GM
|
MilliporeSigma™
5078585GM |
5 g |
Each for $463.00
|
|
|||||
50-785-810GM
|
MilliporeSigma™
50785810GM |
10 g |
Each for $660.00
|
|
|||||
50-785-850GM
|
MilliporeSigma™
50785850GM |
50 g |
Each for $2,387.00
|
|
|||||
Description
PANSORBIN cells are useful as a solid-phase IgG-binding reagent due to the high-affinity interaction of protein A with the Fc domain of IgG. They work better when the antibody is human (IgG1, IgG2, IgG4), rabbit IgG (all isotypes), or mouse (IgG2a, IgG2b, IgG3).
- Activity: ≥2.0 mg human IgG/100mg cells
- Form: Homogeneous, milky suspension
- Formulation: Supplied as a 10% suspension of Staphylococcus aureus cells in PBS, 0.1% NaN3, pH 7.2
- Contaminants: Viable cells: ≤2500 cells/mL
- Storage: Store at 2° to 8°C. Do not freeze
Specifications
PANSORBIN Cells | |
2°C to 8°C |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title