Learn More
Fisherbrand™ Elite Pipette Kit
Kit provides a wide range of pipettes for multiple applications
Description
- Four popular pipette sizes (0.2-2μL, 2-20μL, 20-200μL, 100-1000μL) and stand
- Compatible with universal fit pipet tips
Specifications
Elite | |
Pipette Kit | |
Four pipette sizes (0.2-2uL, 2-20uL, 20-200uL, 100-1000uL) and stand |