Learn More
USA Scientific Inc Electroporation cuvette, 0.4 cm gap
Supplier:  USA Scientific Inc 91046050
Encompass_Preferred
Electroporation cuvette, 0.4 cm gap width, red cap, individually wrapped, sterile, 50/bag
Catalog No. NC9807534