Learn More
Cole-Parmer Oakton by Cole-Parmer Ion-Selective Electrode (ISE), Surfactant, Double-Junction, BNC
Supplier: Cole-Parmer 2750245
These fast and accurate electrodes do not require an external referencethey're easier to use than separate sensing and reference half-cell electrodes. As double-junction combination ion-selective electrodes, they are convenient to use in places where two electrodes won't fit. All electrodes include a 30-mL bottle of reference solution, a reference filling pipette, 1-m cable, BNC connector, and an instruction manual. Use electrodes with a meter that has a resolution of 0.1 mV for the most accurate results.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.