Learn More
Eisco™ WIFI Module
SCI_ED
Allows data to be streamed directly to laptops, tablets, smart phones.
Supplier:  Eisco™ ESPWIFI
Catalog No. S13462
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Description
Description
  • Helpful for collecting field or classroom data without need to interact directly with Base Unit
Operation with WIFI connection to any device
  • Once plugged into Base Unit, Wi-Fi Module broadcasts self-contained Wi-Fi network specific to that Wi-Fi Module
  • No Internet or external network needs to be present in order for this to function
Specifications
Specifications
WiFi Module
Product Suggestions
Product Suggestions