Learn More
Eisco™ Ripple Tank with Screen
SCI_ED
Built-in projection screen for easy display of waves.
Supplier:  Eisco™ PH0768
Catalog No. S05826
Item Discontinued This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative.
Includes: Wave generator unit (vibrator), ripple tank, wave monitor unit, projection mirror, projection screen, footers with adjustment screws, wave dippers, perspex objects, lever arm and support
Description
Description
Full range of accessories included for wave investigations.
  • Electronic strobe and control
  • Reinforced plastic frame with foam beaches to negate reflections
  • Included mirror can be mounted at 45° to project the ripples onto a translucent projection screen on one side of the unit
  • Intense illumination from light source is synchronized with the wave generator to create wave patterns that are easily seen on a lab table
Specifications
Specifications
Ripple Tank
For wave investigations
Wave generator unit (vibrator), ripple tank, wave monitor unit, projection mirror, projection screen, footers with adjustment screws, wave dippers, perspex™ objects, lever arm and support