Learn More
Eisco™ Light Sensor
SCI_ED
Helps students to collect and record light data for experimentation and analysis.
Supplier:  Eisco™ ESP104
Catalog No. S13456
Add to Cart
Requires : Eisco Sensors Base Unit
Description
Description
  • Measures intensity of light in darkest of classrooms to brightest school yards
Specifications:
  • Range: 0-1,000 lx, 0-6,000 lx, 0-150,000 lx
  • Resolution: 1 lx, 6 lx,150 lx
  • Sample Rate: 3,000
Applications:
  • Earth science and weather science studies, behavior of light and light filters, photosynthesis
Specifications
Specifications
Sensor, Light
Earth science and weather science studies, behavior of light and light filters, photosynthesis
Product Suggestions
Product Suggestions