Learn More
Eisco™ Handheld Sensor
Allows students to collect, analyze data directly onto Eisco Sensors Base Unit while on move.
Supplier: Eisco™ ESB101
This item is not returnable.
View return policy
Description
- Can be connected to any Eisco sensor
- Can be used stand alone, tethered to computer, or streaming data wifi to any device
- Built in ambient temperature sensor
- Full color touch screen
- Internal rechargeable battery
Specifications
Sensor, Handheld |