Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Bunsen Burners with Flame Stabilizer

Expanded base prevents easy tipping, even with hoses connected

Supplier:  Eisco™ CH0992LP/RSD

 View more versions of this product

Catalog No. 17-012-825

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.


Description

Description

  • Spaced feet so burner fits comfortably under stands
  • Carry hot burner with ease and safety using finger handle
  • Controls air to fuel ratio via air flow adjustment sleeve; However, it does have flame stabilizer which makes it safer in laboratory setting
  • 16.5cm (6.5 in.) tall
Specifications

Specifications

LPG
4.25 in. (Base)
Flame stabilizer
6 in.
Flame Stabilizer, Handle
1550°C
Safe Continuous Stream of Flammable Gas
Iron
8 mm
16.5 cm
2822°F
4.25 (Base) in.
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit