Learn More
Edwards Fittings for BOC High Vacuum Pumps: Blank Flange
For use with BOC high vacuum pumps
$24.00 - $52.50
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
11-283-16
|
Edwards
C10512368 |
NW16 Blank Flange |
Each for $24.00
|
|
|||||
11-283-17
|
Edwards
C10514368 |
NW25 Blank Flange |
Each for $30.50
|
|
|||||
11-283-18
|
Edwards
C10516368 |
NW40 Blank Flange |
Each for $52.50
|
|
|||||
Description
- Made of aluminum
- Aluminum is ideal for achieving dependable cost-effective performance down to 10-7mbar
Specifications
BOC high vacuum pump | |
Aluminum |