Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Edvotek™ Advanced Placement Laboratory Kit 1: Principles and Practice of Diffusion and Osmosis

Experiments help students develop an understanding of the molecular basis of diffusion and osmosis and its physiological importance.
Supplier: Edvotek™ 281
Includes: instructions, orange indicator dye, violet dye, sucrose, NaCl solution, powdered sucrose, dialysis tubing
Requires :
- 300 to 400mL Beakers† (20 for Part A, 50 for Part B, and 16 for Part C)
- 1, 5, and 10mL Pipets
- Graph paper
- Scales
- Distilled or deionized water
- 10 Thermometers
- Four potatoes
- Cork borers or small kitchen knives
Description
- Analyze how solute size and concentration affect diffusion across semi-permeable membranes and how these processes affect water potential
- Calculate water potential of plant cells
- Class size: 40 students working in groups of 4
Part B: Experiment approximates an osmotic system by demonstrating net flow of water through a dialysis membrane.
Part C: Involves the calculation of potato cell water potential from experimental data.
Order Info
Class size: 40 students working in groups of 4.
Specifications
AP Biology Lab Kit 1: Principles and Practice of Diffusion and Osmosis | |
Low molecular weight indicator dye, high molecular weight violet dye, 1m sucrose, 1m nacl solution, powdered sucrose, dialysis tubing, comprehensive instructions, background material and study questions | |
Room temperature |
Experiments help students develop an understanding of the molecular basis of diffusion and osmosis and its physiological importance, analyze how solute size and concentration affect diffusion across semi-permeable membranes and how these processes affect | |
60 to 90 min. | |
40 Students |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title