Learn More
Malvern Panalytical Edta Peaq-Itc Kit
Supplier: Malvern Panalytical KIT020992
EDTA Test Kit Auto and Manual PEAQ ITC. This kit allows users to test instrument performance using EDTA. Expect material for least 5 runs (with 6 duplicates) from this kit. EDTA Test Kit for Manual PEAQ ITC. Expect material for approximately 30 single runs. This kit will ship with at least 90 days of use before material expiry.