Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eisco™ Economy Steel Tuning Fork
Economy steel tuning forks are designed for sound wave lessons and exploration
Supplier: Eisco™ PH0736G
Description
- Chrome plated steel tuning fork
- Economy priced
- Frequency within ±5%
Specifications
Tuning Fork | |
Chrome-plated Steel | |
Science Experiments | |
480 Hz |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction