Learn More
Cole-Parmer Cole-Parmer StableTemp Economy Gravity Convection Oven, 1.8 cu ft; 120 VAC
Supplier:  Cole-Parmer 5241283
Encompass_Preferred
Gravity convection ensures gentle, natural, air circulation — ideal for sterilization, drying, and other applications requiring subtle airflow. Ovens are programmable for delayed start and stop, and an integrated timer affords the convenience of unattended operation. The dual-wall door opens 180° for greater access to the chamber. Rounded inner chamber corners offer easy cleaning and superior air circulation. Two 1.5" (3.8 cm) diameter vents are located on the top of the unit—adjustable ventilation provides control of inner air and vapor circulation.
Catalog No. NC1426227
Item Discontinued This item has been discontinued and is no longer available. Please reach out to your sales representative.