Learn More
Cederlane ECOLI OK O157 ANTISERA
SDP
Supplier:  Cederlane 44309(SS)
Encompass
OK O157,(No. of tests: 150),(SSI E. COLI OK O SINGLE ANTISERA FOR SLIDE AGGLUTINATION), 3ml vial
Catalog No. NC0168489
Due to product restrictions, please to purchase or view availability for this product.