Learn More
Cygnus Technologies E. coli Host Cell DNA Detection Kit in Tubes
Supplier: Cygnus Technologies D410T
This method utilizes a 2 ml microfuge tube format to measure residual E. coli host cell DNA. The kit includes our proprietary extraction procedure, which isolates residual DNA without the use of iodine or glycogen.The superior co-precipitating reagents do not interfere with qPCR and ensure that pellets do not become dislodged during the extraction procedure. This eliminates the need to centrifuge samples again as well as the need to retest or repeat a run. Cell line-specific E. coli DNA calibrators are included, and upon completion of extraction, samples are reacted with PicoGreen solution. Time to completion is under 2 hours. Each kit supplies enough reagent for the analysis of 50 samples in duplicate.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.