Learn More
California Pacific Lab & Consulting ECO FUNNEL 8IN SYSTEM
SDP
Supplier:  California Pacific Lab & Consulting EF3004GLSYS
Encompass_Preferred
8 in. SAFETY ECO FUNNEL SYSTEM FOR LIQUID WASTE COLLECTION, PREVENTS 99.9% EMISSIONS OF TOXIC FUMES, HDPE, RED LID WITH GASKET THAT LATCHES, REMOVABLE FILTER FOR LARGE PARTICLES, 2000 ML CAPACITY, AIDS IN FIRE PREVENTION. INCLUDES 4 L AMBER GLASS JUG WITH
Catalog No. NC0219500