Learn More
AB Sciex LLC DNA Capillary 100 µm ID x 65 cm
Supplier: AB Sciex LLC 477477
All Capillaries are for use in PA 800 series and P/ACE MDQ Capillary Cartridges for separation and analysis of a variety of samples including pharmaceuticals industrial chemicals chiral drugs nucleic acids amino acids proteins and peptides. Capillaries must be trimmed to optimize speed/resolution.