Learn More
SiliCycle™ E-PAK™ Cartridges, Pilot Scale, SiliaFlash™ Silica Gel F60, 60 - 200 μm, 60 Å
E-PAK™ Cartridges, Pilot Scale, SiliaFlash™ Silica Gel F60, 60 - 200 μm, 60 Å conveniently packaged in a cartridge.
Specifications
Type Cartridge
Particle Size 40 to 63 μm
Pore Size 60 Å
View More Specs
Products 2
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
01-182-138
View Documents
SiliCycle™
PSR10040B1612
12 x 16.5 cm Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
01-182-139
View Documents
SiliCycle™
PSR10040B1625
25 x 16.5 cm Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
Description
Description
  • Narrow particle size distribution
  • Superior resolution
  • Suitable for use with complex matrices
  • Suitable for organic molecules (MW <1000 Dalton)
  • Testing has shown a good correlation between the performance of lab, pilot, and commercial size cartridges.
Specifications
Specifications
Cartridge
40 to 63 μm
60 Å