Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SiliCycle™ E-PAK™ Cartridges, Lab Scale, SiliaFlash™ Irregular Silica Gel F60, 40 to 63 μm, 60 Å
E-PAK™ Cartridges, Lab Scale, SiliaFlash™ Irregular Silica Gel F60, 40 - 63 μm, 60 Å is conveniently packaged in a cartridge. E-PAK™ lab cartridges are generally used for processing samples of 50 mL or more.
$161.20 - $1395.00
Specifications
Type | Lab Scale E-PAK Cartridge |
---|---|
For Use With (Application) | Removal of various polar compounds |
Dimensions (L x W) | 1 x 5 cm |
Particle Size | 40 to 63 μm |
Pore Size | 60 Å |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
01-182-075
|
SiliCycle™
LSR10030B51 |
1 Pk. |
Each for $161.20
|
|
|||||
01-182-077
|
SiliCycle™
LSR10030B513 |
3 Pk. |
Each for $457.00
|
|
|||||
01-182-078
|
SiliCycle™
LSR10030B515 |
5 Pk. |
Each for $737.50
|
|
|||||
01-182-076
|
SiliCycle™
LSR10030B5110 |
10 Pk. |
Each for $1,395.00
|
|
|||||
Description
- Narrow particle size distribution
- Superior resolution
- Suitable for use with complex matrices
- Suitable for organic molecules (MW <1000 Dalton)
- E-PAK™ lab cartridges are generally used for processing samples of 50 mL or more.
- Testing has shown a good correlation between the performance of lab, pilot, and commercial size cartridges.
- Lab cartridges can be operated with a peristaltic pump or a small pressure vessel.
Specifications
Lab Scale E-PAK Cartridge | |
1 x 5 cm | |
60 Å | |
8 g |
Removal of various polar compounds | |
40 to 63 μm | |
Bare Silica (Silicon Dioxyde, SiO2) |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title