Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M™ E-A-R™ E-Z-Fit™ Ear Plugs
Click to view available options
Model:
312-1208
312-1222
Corded:
Corded
Uncorded
Description
- Ideal for smaller ear canals
- Poly bag keeps each pair of ear plugs clean before use
- Test compatible with 3M™ E-A-Rfit™ Validation System
Compliance
3M™ ear plugs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Specifications
ANSI S3.1974 | Yes |
Cord Color | Blue |
Cord Material | Vinyl |
CSA Z94.2 | Class AL |
Size | Small |
For Use With (Application) | Assembly, Blasting, Cleaning, Demolition, Electrical, Facility Maintenance, Grinding, Machine Operations, Painting, Sanding, Welding |
High Visibility | Yes |
Model | 312-1222 |
Packaging Type | Poly bag |
Pairs per Package | 200 Pairs/Pk. |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction