Learn More
CTL Scientific Supply Corp DUOTEST PH 9.5-14 DISP,PK/3 RL
Supplier: CTL Scientific Supply Corp 903063
Duotest indicator pH paper 9.5-14.0, pack of 3 rolls in dispensers with color charts, papers with two complimentary indicator zones on one strip. Gradation of scale: 9.5 - 10.0 - 10.5 - 11.0 - 11.5 - 12.0 - 12.5 - 13.0 - 13.5 - 14.0