Learn More
Thermo Scientific DUAL SPRAY CHAMBER
Supplier: Thermo Scientific 842312051411
Spray Chamber; Standard glass cyclonic spraychamber provided with all iCAP 6000; Series Spectrometers.; Requires an appropriate adaptor dependent on whether a Radial or Duo; instrument is used