Learn More
Andwin Scientific DUAL PEEL 9IN TUBNG 100FT ROLL
Supplier: Andwin Scientific T90009
V. Mueller DualPeel Gas/Steam Sterilization Tubing, Blue-tinted plastic and surgical-grade paper combination includes two chemical process indicators. Cut tubing to the length needed, insert item, and heat-seal the open ends.