Learn More
Applied Biosystems™ DS-32 Matrix Standard Set [5FAM™, JOE™, NED™ and ROX™ dyes] for the 310 Genetic Analyzer
Software for these instruments uses the multicomponent matrix to automatically analyze the five different colored fluorescent dye-labeled samples.
Manufacturer: Applied Biosystems™ 4312131
Includes: Contains dye-labeled oligonucleotides used to generate the multicomponent matrix required for the AmpF/STR® kit.
Description
The matrix standard st is used to generate the 'multicomponent matrix' required when analyzing DNA fragments on the 310 genetic anlayzer.Order Info
Kit
Specifications
Contains dye-labeled oligonucleotides used to generate the multicomponent matrix required for the AmpF⁄STR™ kit. Store at 2 - 8°C. Do not freeze. | |
Sequencing | |
5FAM™, JOE™, NED™, ROX™ | |
1 Kit |
310 Genetic Analyzer | |
FAM, JOE, NED, ROX | |
DS-32 Matrix Standard Set | |
Wet Ice |