Learn More
Utak DRUG FREE URINE LIQ 1GAL
Supplier: Utak 88121CDF(L)
Certified drug-free urine can be utilized to prepare controls for drugs or other compounds which are not commercially available. 1 gallon.