Learn More
Drucker™ 6-Series SmartView Platform
Tilts centrifuge to better display rotor chamber. Drucker 6-Series SmartView Platform provides healthcare professionals with clear view while loading and unloading specimens. With the SmartView platform, centrifuges can easily be monitored from across the lab.
Supplier: Drucker™ 00079009001
Description
- Centrifuges fit snugly into SmartView platform
- Four rubber feet secure platform and quiet vibrations
- Facilitates viewing in cramped settings (under counter, in car for visiting nurses)
- Leaves centrifugation unaffected as centrifugal forces act on axis of tubes
- Compatible with models 614, 642, and any other model in 600 series
- Fits many other nameplate centrifuges (LabCorp, Quest Diagnostics, DaVita and many others)
Specifications
Color: White | |
Models 614B, 642B, 642E, 642VES | |
Platform | |
2 Years |