Learn More
United States Biological Corporation DROSOPHILA GRADE AGAR 2.5KG
Supplier: United States Biological Corporation A09402.5KG
Gelidium based Drosophila agar is an inexpensive growth medium especially suited to the growth and nutrient requirements of Drosophila without the expense of bacteriological grades. This is a food grade agar which offers excellent clarity. Appearance: White to light tan, homogenous, free flowing powder. Melting Point (1.5%): 82-88°C pH (1.5%): 6.0-8.0 Water: ≤11% Ash: ≤6.5% Gel Point (1.5%): 34-38°C Gel Strength (1.5%): 600-1100g/cm2 Identification A: Passes Identification B: Passes Foreign Insoluble Matter: ≤10% Foreign Organic Matter: ≤1% Foreign Starch: None Detected Gelatin: None Detected Water Absorption: ≤75ml Acid-insoluble Ash: ≤0.5% Arsenic: ≤0.0003% Lead: ≤0.001% Heavy Metals (Pb): ≤0.004% Microbiological Analysis: Total Plate Count: ≤500cfu/g Coliforms: ≤3cfu/g E. coli: 0cfu/g Salmonella: 0cfu/g Yeast and Mold: ≤200cfu/g
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.