Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dried Aluminum Hydroxide Gel, Powder, USP, 76.5%, Spectrum™ Chemical
SDP

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Pail
21645-51-2
AlH6O3
81.03
MXRIRQGCELJRSN-UHFFFAOYSA-N
alumanetriol
O.O.O.[Al]
21645-51-2
AlH6O3
81.03
MXRIRQGCELJRSN-UHFFFAOYSA-N
alumanetriol
O.O.O.[Al]

AL226, 21645-51-2, Al(OH)3

Spectrum™ Chemical Dried Aluminum Hydroxide Gel, Powder, USP is used as an antacid. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
Quantity 50 kg
Percent Purity 76.5%
Grade USP
Packaging Fiber Drum
pH Range 10
Arsenic (As) 8 ppm
Sulfate 0.6%
Chloride 0.85%