Learn More
Thermo Scientific™ DRI™ Ethyl Glucuronide (EtG) Drugs of Abuse Control Sets
Save time while maintaining accuracy with these liquid, ready-to-use control sets, which are for Criminal Justice and Forensic Use Only and are for sale in the U.S.A. only.
$414.00 - $429.00
Specifications
Content And Storage | 2°C to 8°C |
---|---|
Detectable Analytes | Ethyl Glucuronide (EtG) |
Sample Type | Urine |
Type | Control |
Quantity | 1 x 25 mL |
Alert:
For Criminal Justice and Forensic use only.
Catalog Number | Mfr. No. | Concentration | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Concentration | Price | Quantity & Availability | |||||
23-278-362
|
Thermo Scientific™
10015934 |
375 ng/mL |
Each for $437.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
23-278-364
|
Thermo Scientific™
10015936 |
625 ng/mL |
Each for $429.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
23-278-365
|
Thermo Scientific™
10015937 |
750 ng/mL |
Each for $414.00
|
|
|||||
23-278-367
|
Thermo Scientific™
10015939 |
1250 ng/mL |
Each for $429.00
|
|
|||||
Description
Streamline your testing process while maintaining high analytical accuracy by utilizing Thermo Scientific™ DRI™ Ethyl Glucuronide (EtG) Drugs of Abuse Control Sets along with our Thermo Scientific DRI Ethyl Glucuronide (EtG) Drugs of Abuse Assays and Calibrators. These control sets are for Criminal Justice and Forensic Use Only and are for sale in the U.S.A. only.
- Specific detection of Ethyl Glucuronide (EtG)
- Qualitative or semi-quantitative modes
- Excellent correlation to LC-MS/MS
- Applications for an array of clinical chemistry analyzers
Specifications
2°C to 8°C | |
Urine | |
1 x 25 mL |
Ethyl Glucuronide (EtG) | |
Control |
For sale in the U.S.A. only.
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.