Learn More
Thermo Scientific™ DRI™ Alcohol Negative Drugs of Abuse Calibrators
Eliminate time-consuming steps in reagent preparation while still achieving high analytical accuracy.
$83.00 - $402.00
Specifications
Description | DRI Alcohol Negative Drugs of Abuse Calibrators |
---|---|
Concentration | 0 ng/mL |
Detectable Analytes | Alcohol |
Sample Type | Plasma, Serum, Urine |
Type | Calibrator |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
23-278-318
|
Thermo Scientific™
0311 |
1 x 5 mL |
Each for $83.00
|
|
|||||
23-278-410
|
Thermo Scientific™
1405 |
1 x 25 mL |
Each for $402.00
|
|
|||||
Description
Eliminate time-consuming steps in reagent preparation while still achieving high analytical accuracy. Ready-to-use in liquid form, Thermo Scientific™ DRI™ Alcohol Negative Drugs of Abuse Calibrators have been field-tested on a wide range of analyzers. They are enthusiastically accepted in laboratories where cost control and optimal productivity are key issues.
- Qualitative or semi-quantitative options for results
- Excellent correlation to GC/MS
- Applications for an array of clinical chemistry analyzers
- Variety of calibrators and multiconstituent control sets
Specifications
DRI Alcohol Negative Drugs of Abuse Calibrators | |
Alcohol | |
Calibrator |
0 ng/mL | |
Plasma, Serum, Urine | |
1 Bottle |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.