Learn More
Thermo Scientific™ DRI™ Ethyl Alcohol Drugs of Abuse Calibrators
Choose Thermo Scientific™ DRI™ Ethyl Alcohol Drugs of Abuse Calibrators to deliver high analytical accuracy while eliminating time-consuming steps in reagent preparation.
$86.00 - $408.00
Specifications
Content And Storage | 2°C to 8°C |
---|---|
Description | DRI Alcohol Drugs of Abuse Calibrators |
Concentration | 100 mg/dL |
Detectable Analytes | Alcohol |
Sample Type | Plasma, Serum, Urine |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
23-278-316
|
Thermo Scientific™
0241 |
1 x 5 mL |
Each for $86.00
|
|
|||||
23-278-411
|
Thermo Scientific™
1406 |
1 x 25 mL |
Each for $408.00
|
|
|||||
Description
Choose Thermo Scientific™ DRI™ Ethyl Alcohol Drugs of Abuse Calibrators to deliver high analytical accuracy while eliminating time-consuming steps in reagent preparation. Ready-to-use in liquid form, these assays have been field-tested on a wide range of analyzers. They are enthusiastically accepted in laboratories where cost control and optimal productivity are key issues.
- Qualitative or semi-quantitative options for results
- Excellent correlation to GC/MS
- Applications for an array of clinical chemistry analyzers
- Variety of calibrators and multiconstituent control sets
Specifications
2°C to 8°C | |
100 mg/dL | |
Plasma, Serum, Urine | |
1 Bottle |
DRI Alcohol Drugs of Abuse Calibrators | |
Alcohol | |
Calibrator |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.