Learn More
Cole-Parmer Honeywell 10" Chart Recorder with digital display, 1 channel
Supplier: Cole-Parmer 8066120
Recorders come built with a stepper motor for precise operation. The molded housing has a gasketed door and acrylic window to meet NEMA 3 requirements; panel on surface mount.