Learn More
Metrohm USA DOSING UNIT 807-5ML 6.3032.150
Supplier: Metrohm USA 63032150
Dosing unit with integrated data chip with 5 mL glass cylinder and light protection, mountable on reagent bottle with ISO/DIN GL45 thread. FEP tubing connection, anti-diffusion buret tip.