Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DMPO, Tocris Bioscience™
Click to view available options
Quantity:
50mg
Description
Water soluble nitric oxide spin trap; allows the measurement of oxygen-centered free radicals in biological systems at room temperature using electron spin resonance (ESR). Has a high reaction rate constant for superoxide and hydroxyl radicals, and distinguishes simultaneously among a variety of important biologically generated free radicals.
Specifications
Quantity | 50mg |
Formula Weight | 113.16 |
Percent Purity | >98% |
Chemical Name or Material | DMPO |
Safety and Handling
Recommended Storage : Store at -20°C