Learn More
Anton Paar DMA 35 Ampere Portable density meter for battery acid
Supplier:  Anton Paar 226130
Encompass_Preferred
DMA 35 Ampere is a portable density meter for measuring the specific gravity of battery acids during production and maintenance. Only 2 mL of sample are required for a measurement, performed based on the oscillating U-tube technology. If the measuring cell nevertheless suffers from cell rupture, you benefit from the unique patented connection of the measuring cell, which allows replacement. An RFID interface for quick sample identification and a Bluetooth interface for convenient data handling in the field are integral parts of the instrument. DMA 35 Ampere has protection class IP54 and high chemical resistance for use in harsh environments.Measuring range from 0 g/cm3 up to 3 g/cm3Accuracy of Density: 0.001 g/cm3Accuracy of Displayed Temperature: 0.2°CAmbient temperature suitable for -10°C up to +50°CIncludes Carrying Case.
Catalog No. NC1818211