Learn More
Disulfoton, Reference Material, MilliporeSigma™ Supelco™
This reference material (RM) is produced and certified in accordance with ISO/IEC 17025. This RM is traceable to primary material from an NMI, e.g. NIST or NMIJ. Certified content incl. uncertainty and expiry date are given on the certificate.
Supplier: MilliporeSigma Supelco 49784100MG
Specifications
298-04-4 | |
C8H19O2PS3 | |
274.40 | |
Reference Material | |
133°C |
C8H19O2PS3 | |
MFCD00036234 | |
≥98% (GC) | |
100 mg |
Safety and Handling
H300 + H310 - H410
ShelfLife : Limited shelf life, expiry date on the label
RTECSNumber : TD9275000
Recommended Storage : 2°C to 8°C
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.